Anti-CYP3A4 Rabbit Polyclonal Antibody

Lieferant: US Biological
125584
USBI125584EA 662 EUR
USBI125584
Anti-CYP3A4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP3A4 (cytochrome P450, family 3, subfamily A, polypeptide 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

TECHNISCHE DATEN

Antigen-Symbol CYP3A4
Antigen-Name cytochrome P450, family 3, subfamily A, polypeptide 4
Environmentally Preferable
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human
Wirt Rabbit
Isotyp IgG
Western blot Yes
Form liquid
Reinigung purified by Protein A chromatography
Lagerungspuffer PBS, pH 7,2.
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR