Anti-CYP4V2 Rabbit Polyclonal Antibody

Lieferant: US Biological
125597
USBI125597EA 696 EUR
USBI125597
Anti-CYP4V2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP4V2 (cytochrome P450, family 4, subfamily V, polypeptide 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR