Anti-DDT Mouse Monoclonal Antibody [clone: 1G1]

Lieferant: US Biological
125720
USBI125720EA 623 EUR
USBI125720
Anti-DDT Mouse Monoclonal Antibody [clone: 1G1]
Antikörper
Type: Primary
Antigen: DDT (D-dopachrome tautomerase)
Clonality: monoclonal
Clone: 1G1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR