Anti-DDX42 Mouse Polyclonal Antibody

Lieferant: US Biological
125733
USBI125733EA 623 EUR
USBI125733
Anti-DDX42 Mouse Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: DDX42 (DEAD (Asp-Glu-Ala-Asp) box helicase 42)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR