Anti-DPH5 Mouse Polyclonal Antibody

Lieferant: US Biological
125998
USBI125998EA 623 EUR
USBI125998
Anti-DPH5 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DPH5 (diphthamide biosynthesis 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR