Anti-DUSP14 Mouse Monoclonal Antibody [clone: 4B5-E6]
Lieferant: US Biological
126057
USBI126057EA 623 EUR
USBI126057
Anti-DUSP14 Mouse Monoclonal Antibody [clone: 4B5-E6]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: DUSP14 (dual specificity phosphatase 14)
Clonality: monoclonal
Clone: 4B5-E6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human