Anti-EMD Mouse Monoclonal Antibody [clone: 3B9]

Lieferant: US Biological
126299
USBI126299EA 623 EUR
USBI126299
Anti-EMD Mouse Monoclonal Antibody [clone: 3B9]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 1 ?g/ml

Type: Primary
Antigen: EMD (emerin)
Clonality: monoclonal
Clone: 3B9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR