Anti-EOMES Mouse Monoclonal Antibody [clone: 3F1]

Lieferant: US Biological
126354
USBI126354EA 482 EUR
USBI126354
Anti-EOMES Mouse Monoclonal Antibody [clone: 3F1]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: EOMES (eomesodermin)
Clonality: monoclonal
Clone: 3F1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR