Anti-ERAP1 Mouse Monoclonal Antibody [clone: 4H8]

Lieferant: US Biological
126417
USBI126417EA 655 EUR
USBI126417
Anti-ERAP1 Mouse Monoclonal Antibody [clone: 4H8]
Antikörper
Type: Primary
Antigen: ERAP1 (endoplasmic reticulum aminopeptidase 1)
Clonality: monoclonal
Clone: 4H8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR