Anti-EXO1 Rabbit Polyclonal Antibody

Lieferant: US Biological
126498
USBI126498EA 662 EUR
USBI126498
Anti-EXO1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EXO1 (exonuclease 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR