Anti-F13A1 Mouse Monoclonal Antibody [clone: M1]

Lieferant: US Biological
126537
USBI126537EA 623 EUR
USBI126537
Anti-F13A1 Mouse Monoclonal Antibody [clone: M1]
Antikörper
Type: Primary
Antigen: F13A1 (coagulation factor XIII, A1 polypeptide)
Clonality: monoclonal
Clone: M1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR