Anti-FABP4 Rabbit Polyclonal Antibody

Lieferant: US Biological
126551
USBI126551EA 696 EUR
USBI126551
Anti-FABP4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FABP4 (fatty acid binding protein 4, adipocyte)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR