Anti-FKBP14 Mouse Polyclonal Antibody

Lieferant: US Biological
126828
USBI126828EA 655 EUR
USBI126828
Anti-FKBP14 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FKBP14 (FK506 binding protein 14, 22 kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR