Anti-AIDA Mouse Monoclonal Antibody [clone: 2E6-1C11]

Lieferant: US Biological
126849
USBI126849EA 623 EUR
USBI126849
Anti-AIDA Mouse Monoclonal Antibody [clone: 2E6-1C11]
Antikörper
Recommended dilution: Immunofluorescence: 60 ?g/ml

Type: Primary
Antigen: AIDA (axin interactor, dorsalization associated)
Clonality: monoclonal
Clone: 2E6-1C11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR