Anti-FLT3LG Mouse Monoclonal Antibody [clone: 4C4]

Lieferant: US Biological
126897
USBI126897EA 655 EUR
USBI126897
Anti-FLT3LG Mouse Monoclonal Antibody [clone: 4C4]
Antikörper
Type: Primary
Antigen: FLT3LG (fms-related tyrosine kinase 3 ligand)
Clonality: monoclonal
Clone: 4C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR