Anti-GART Mouse Polyclonal Antibody

Lieferant: US Biological
127169
USBI127169EA 623 EUR
USBI127169
Anti-GART Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GART (phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR