Anti-GNE Rabbit Polyclonal Antibody

Lieferant: US Biological
127424
USBI127424EA 662 EUR
USBI127424
Anti-GNE Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GNE (glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR