Anti-GRIN2C Mouse Polyclonal Antibody

Lieferant: US Biological
127544
USBI127544EA 623 EUR
USBI127544
Anti-GRIN2C Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GRIN2C (glutamate receptor, ionotropic, N-methyl D-aspartate 2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR