Anti-GSTZ1 Rabbit Polyclonal Antibody

Lieferant: US Biological
127620
USBI127620EA 696 EUR
USBI127620
Anti-GSTZ1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GSTZ1 (glutathione S-transferase zeta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR