Anti-H2AFY2 Rabbit Polyclonal Antibody

Lieferant: US Biological
127695
USBI127695EA 696 EUR
USBI127695
Anti-H2AFY2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: H2AFY2 (H2A histone family, member Y2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR