Anti-HDAC9 Mouse Monoclonal Antibody [clone: 2G10]

Lieferant: US Biological
127776
USBI127776EA 655 EUR
USBI127776
Anti-HDAC9 Mouse Monoclonal Antibody [clone: 2G10]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: HDAC9 (histone deacetylase 9)
Clonality: monoclonal
Clone: 2G10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR