Anti-HERPUD1 Rabbit Polyclonal Antibody

Lieferant: US Biological
127808
USBI127808EA 662 EUR
USBI127808
Anti-HERPUD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HERPUD1 (homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-like domain member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR