Anti-HERPUD1 Mouse Monoclonal Antibody [clone: 2G7]
Lieferant: US Biological
127809
USBI127809EA 623 EUR
USBI127809
Anti-HERPUD1 Mouse Monoclonal Antibody [clone: 2G7]
Antikörper
Recommended dilution: Immunochemistry: 3?g/ml
Type: Primary
Antigen: HERPUD1 (homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-like domain member 1)
Clonality: monoclonal
Clone: 2G7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human