Anti-HMGCR Rabbit Polyclonal Antibody

Lieferant: US Biological
127959
USBI127959EA 662 EUR
USBI127959
Anti-HMGCR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HMGCR (3-hydroxy-3-methylglutaryl-CoA reductase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR