Anti-HOXA5 Mouse Polyclonal Antibody

Lieferant: US Biological
128011
USBI128011EA 655 EUR
USBI128011
Anti-HOXA5 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HOXA5 (homeobox A5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR