Anti-HPD Mouse Monoclonal Antibody [clone: 2F3]

Lieferant: US Biological
128056
USBI128056EA 623 EUR
USBI128056
Anti-HPD Mouse Monoclonal Antibody [clone: 2F3]
Antikörper
Type: Primary
Antigen: HPD (4-hydroxyphenylpyruvate dioxygenase)
Clonality: monoclonal
Clone: 2F3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR