Anti-ISG20 Rabbit Polyclonal Antibody

Lieferant: US Biological
128603
USBI128603EA 696 EUR
USBI128603
Anti-ISG20 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ISG20 (interferon stimulated exonuclease gene 20kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR