Anti-Kappa Light Chain Rabbit Polyclonal Antibody

Lieferant: US Biological
128719
USBI128719EA 662 EUR
USBI128719
Anti-Kappa Light Chain Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Kappa Light Chain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR