Anti-KIRREL2 Mouse Monoclonal Antibody [clone: 2B9-1D3]

Lieferant: US Biological
128839
USBI128839EA 655 EUR
USBI128839
Anti-KIRREL2 Mouse Monoclonal Antibody [clone: 2B9-1D3]
Antikörper
Type: Primary
Antigen: KIRREL2 (kin of IRRE like 2 (Drosophila))
Clonality: monoclonal
Clone: 2B9-1D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR