Anti-LAMC1 Mouse Monoclonal Antibody [clone: M1]

Lieferant: US Biological
128992
USBI128992EA 623 EUR
USBI128992
Anti-LAMC1 Mouse Monoclonal Antibody [clone: M1]
Antikörper
Type: Primary
Antigen: LAMC1 (laminin, gamma 1 (formerly LAMB2))
Clonality: monoclonal
Clone: M1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR