Anti-LCN2 Mouse Polyclonal Antibody

Lieferant: US Biological
129030
USBI129030EA 623 EUR
USBI129030
Anti-LCN2 Mouse Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR