Anti-LGMN Mouse Monoclonal Antibody [clone: M1]
Lieferant: US Biological
129076
USBI129076EA 623 EUR
USBI129076
Anti-LGMN Mouse Monoclonal Antibody [clone: M1]
Antikörper
Type: Primary
Antigen: LGMN (legumain)
Clonality: monoclonal
Clone: M1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human