Anti-MARCH2 Mouse Polyclonal Antibody

Lieferant: US Biological
129427
USBI129427EA 623 EUR
USBI129427
Anti-MARCH2 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MARCH2 (membrane-associated ring finger (C3HC4) 2, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR