Anti-MLH1 Mouse Monoclonal Antibody [clone: M1]
Lieferant: US Biological
129719
USBI129719EA 623 EUR
USBI129719
Anti-MLH1 Mouse Monoclonal Antibody [clone: M1]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 1 ?g/ml
Type: Primary
Antigen: MLH1 (mutL homolog 1)
Clonality: monoclonal
Clone: M1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human