Anti-MUSK Mouse Monoclonal Antibody [clone: 2H6]

Lieferant: US Biological
130014
USBI130014EA 623 EUR
USBI130014
Anti-MUSK Mouse Monoclonal Antibody [clone: 2H6]
Antikörper
Recommended dilution: Immunochemistry: 1 ?g/ml

Type: Primary
Antigen: MUSK (muscle, skeletal, receptor tyrosine kinase)
Clonality: monoclonal
Clone: 2H6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR