Anti-NBN Mouse Monoclonal Antibody [clone: 3E4]

Lieferant: US Biological
130364
USBI130364EA 623 EUR
USBI130364
Anti-NBN Mouse Monoclonal Antibody [clone: 3E4]
Antikörper
Type: Primary
Antigen: NBN (nibrin)
Clonality: monoclonal
Clone: 30000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR