Anti-NISCH Mouse Monoclonal Antibody [clone: 2C8]

Lieferant: US Biological
130383
USBI130383EA 623 EUR
USBI130383
Anti-NISCH Mouse Monoclonal Antibody [clone: 2C8]
Antikörper
Type: Primary
Antigen: NISCH (nischarin)
Clonality: monoclonal
Clone: 2C8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR