Anti-p40 phox Rabbit Polyclonal Antibody

Lieferant: US Biological
130798
USBI130798EA 696 EUR
USBI130798
Anti-p40 phox Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: p40 phox
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR