Anti-PGM2L1 Mouse Polyclonal Antibody

Lieferant: US Biological
131219
USBI131219EA 623 EUR
USBI131219
Anti-PGM2L1 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PGM2L1 (phosphoglucomutase 2-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR