Anti-Phospholipase A2, Secretory, Group IB Rabbit Polyclonal Antibody

Lieferant: US Biological
131288
USBI131288EA 696 EUR
USBI131288
Anti-Phospholipase A2, Secretory, Group IB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Phospholipase A2, Secretory, Group IB
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR