Anti-PI15 Mouse Monoclonal Antibody [clone: 3B5]

Lieferant: US Biological
131298
USBI131298EA 655 EUR
USBI131298
Anti-PI15 Mouse Monoclonal Antibody [clone: 3B5]
Antikörper
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: PI15 (peptidase inhibitor 15)
Clonality: monoclonal
Clone: 3B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR