Anti-PIR Rabbit Polyclonal Antibody

Lieferant: US Biological
131370
USBI131370EA 662 EUR
USBI131370
Anti-PIR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PIR (pirin (iron-binding nuclear protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR