Anti-PPIL4 Mouse Polyclonal Antibody

Lieferant: US Biological
131662
USBI131662EA 655 EUR
USBI131662
Anti-PPIL4 Mouse Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: PPIL4 (peptidylprolyl isomerase (cyclophilin)-like 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR