Anti-PSMB2 Mouse Monoclonal Antibody [clone: M1]

Lieferant: US Biological
131951
USBI131951EA 623 EUR
USBI131951
Anti-PSMB2 Mouse Monoclonal Antibody [clone: M1]
Antikörper
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: PSMB2 (proteasome (prosome, macropain) subunit, beta type, 2)
Clonality: monoclonal
Clone: M1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR