Anti-Retinol Dehydrogenase 12 Rabbit Polyclonal Antibody

Lieferant: US Biological
132441
USBI132441EA 696 EUR
USBI132441
Anti-Retinol Dehydrogenase 12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Retinol Dehydrogenase 12
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR