Anti-Ring Finger Protein 40 Mouse Monoclonal Antibody [clone: 1C1]

Lieferant: US Biological
132589
USBI132589EA 655 EUR
USBI132589
Anti-Ring Finger Protein 40 Mouse Monoclonal Antibody [clone: 1C1]
Antikörper
Type: Primary
Antigen: Ring Finger Protein 40
Clonality: monoclonal
Clone: 1C1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR