Anti-RLIM Rabbit Polyclonal Antibody

Lieferant: US Biological
132639
USBI132639EA 662 EUR
USBI132639
Anti-RLIM Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RLIM (ring finger protein, LIM domain interacting)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR