Anti-RING Finger Protein 20 Rabbit Polyclonal Antibody
Lieferant: US Biological
132684
USBI132684EA 696 EUR
USBI132684
Anti-RING Finger Protein 20 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RING Finger Protein 20
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human