Anti-MST4 Mouse Monoclonal Antibody [clone: 2G6]
Lieferant: US Biological
132748
USBI132748EA 655 EUR
USBI132748
Anti-MST4 Mouse Monoclonal Antibody [clone: 2G6]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: MST4
Clonality: monoclonal
Clone: 2G6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human