Anti-RPAP3 Rabbit Polyclonal Antibody

Lieferant: US Biological
132758
USBI132758EA 696 EUR
USBI132758
Anti-RPAP3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPAP3 (RNA polymerase II associated protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR